2,4,6-Trimethylbenzylchlorid
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-(Chlormethyl)-2,4,6-trimethylbenzol, α-Chlor-1,2,4,6-tetramethylbenzol
SB01359.1
SB01359.10
SB01359.50
MAYBSB01359.1EA
0
EUR
MAYBSB01359.1
MAYBSB01359.10
MAYBSB01359.50
2,4,6-Trimethylbenzylchlorid
2,4,6-Trimethylbenzylchlorid
Formel:
C₁₀H₁₃Cl Molecular Weight: 168,67 g/mol Siedepunkt: 245 °C (1013 hPa) Schmelzpunkt: 34…38 °C |
MDL:
MFCD00000901 CAS-Nummer: 1585-16-6 EINECS: 216-440-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...