5-Brom-o-kresol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
5-Brom-2-methylphenol, 4-Brom-2-hydroxytoluol, 1-Brom-3-hydroxy-4-methylbenzol
722480-10G
722480-1G
SIAL722480-10GEA
105
EUR
SIAL722480-10G
SIAL722480-1G
5-Brom-o-kresol, Sigma-Aldrich®
5-Brom-o-kresol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol |
MDL:
MFCD08061906 CAS-Nummer: 36138-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...