2-Fluor-5-toluoylchlorid
Lieferant: CHEMPUR
Synonyme:
2-Fluor-5-methylbenzoylchlorid, 2-Fluor-5-tolylsäurechlorid
FL13847.POR
013847-1G
CHMPFL13847.POREA
0
EUR
CHMPFL13847.POR
FLUO013847-1G
2-Fluor-5-toluoylchlorid
2-Fluor-5-toluoylchlorid
Formel:
C₈H₆ClFO Molecular Weight: 172.59 g/mol |
MDL:
MFCD04973776 CAS-Nummer: 135564-61-3 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...