Kalium 99,95 Metals basis, Solid

Lieferant: Thermo Fisher Scientific
Gefahr


breakseal ampoule

45842.03 45842.06
45842.03EA 166 EUR
45842.03 45842.06
Kalium 99,95 Metals basis, Solid
Potassium

Purity: 99.95% CAS No.: 7440-09-7 Physical Form: Sealed in breakseal ampoules Odour: Odourless UN No.: 2257 MDL No.: MFCD00133776

Formel: K
Molecular Weight: 39.1 g/mol
Siedepunkt: 760
Schmelzpunkt: 63
Dichte: 0.86 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00133776
CAS-Nummer: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3 I
Merck Index: 12,07763

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR