1-Brom-3-fluorpropan
Lieferant: SIA ENAMINE
EN300107714.1G
EN300-107714-1
ENAMEN300107714.1GEA
0
EUR
ENAMEN300107714.1G
ENAMEN300-107714-1
1-Brom-3-fluorpropan
1-Bromo-3-fluoropropane
Formel:
BrH₂CCH₂CH₂F Molecular Weight: 140,98 g/mol Siedepunkt: 99 °C (1013 hPa) Dichte: 1,542 g/cm³ (25 °C) |
MDL:
MFCD00039287 CAS-Nummer: 352-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...