2'-Chlor-5'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

005704-1G 005704-5G
FLUO005704-1GEA 51.8 EUR
FLUO005704-1G FLUO005704-5G
2'-Chlor-5'-(trifluormethyl)acetophenon
2'-Chlor-5'-(trifluormethyl)acetophenon
Formel: C₉H₆ClF₃O
Molecular Weight: 222.59 g/mol
MDL: MFCD00045092
CAS-Nummer: 71648-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR