3-Amino-4-brom-1H-pyrazol

Lieferant: CHEMPUR

Synonyme: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

GK00248.1 GK00248.10
CHMPGK00248.1EA 0 EUR
CHMPGK00248.1 CHMPGK00248.10
3-Amino-4-brom-1H-pyrazol
3-Amino-4-bromo-1H-pyrazole
Formel: C₃H₄BrN₃
Molecular Weight: 161.99 g/mol
MDL: MFCD00082728
CAS-Nummer: 16461-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR