5-Amino-1-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

AM-1614-5G AM-1614-25G AM-1614-1G
COBBAM-1614-5GEA 65.7 EUR
COBBAM-1614-5G COBBAM-1614-25G COBBAM-1614-1G
5-Amino-1-methyl-1H-pyrazol
5-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
Molecular Weight: 97.12 g/mol
MDL: MFCD00068156
CAS-Nummer: 1192-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR