2-(Propylthio)nicotinamid
Lieferant: Thermo Scientific
Synonyme:
2-(n-Propylthio)nicotinamide
BTB06388.1
BTB06388.10
BTB06388.50
MAYBBTB06388.1EA
0
EUR
MAYBBTB06388.1
MAYBBTB06388.10
MAYBBTB06388.50
2-(Propylthio)nicotinamid
2-(Propylthio)nicotinamid
Formel:
C₉H₁₂N₂OS Molecular Weight: 196,27 g/mol |
MDL:
MFCD00052904 CAS-Nummer: 175135-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...