5,7-Difluoroxindol

Lieferant: FLUOROCHEM

010572-1G 010572-2.5G 010572-5G
FLUO010572-1GEA 30.3 EUR
FLUO010572-1G FLUO010572-2.5G FLUO010572-5G
5,7-Difluoroxindol
5,7-Difluoroxindol
Formel: C₈H₅F₂NO
MDL: MFCD01075219
CAS-Nummer: 247564-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR