1-Brom-4-(trifluormethylthio)benzol

Lieferant: CHEMPUR

Synonyme: 4-(Trifluoromethylthio)bromobenzene, 4-Bromophenyl trifluoromethyl sulphide

FL1482.5 FL1482.25
CHMPFL1482.5EA 0 EUR
CHMPFL1482.5 CHMPFL1482.25
1-Brom-4-(trifluormethylthio)benzol
1-Bromo-4-(trifluoromethylthio)benzene
Formel: C₇H₄BrF₃S
Molecular Weight: 257,07 g/mol
MDL: MFCD00040840
CAS-Nummer: 333-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR