2,6-Chinolindiol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Hydroxyquinolin-2(1H)-one, 2,6-Dihydroxyquinoline

PY-1182-5G PY-1182-1G PY-1182-25G
COBBPY-1182-5GEA 51.5 EUR
COBBPY-1182-5G COBBPY-1182-1G COBBPY-1182-25G
2,6-Chinolindiol
2,6-Chinolindiol
Formel: C₉H₇NO₂
Molecular Weight: 161.1600 g/mol
MDL: MFCD00239446
CAS-Nummer: 19315-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR