6-Brom-3-formylchromon

Lieferant: CHEMPUR

Synonyme: 6-Bromo-4-oxo-4H-chromene-3-carbaldehyde

RJC01039.250 RJC01039.1
CHMPRJC01039.250EA 0 EUR
CHMPRJC01039.250 MAYBRJC01039.1
6-Brom-3-formylchromon
6-Brom-3-formylchromon
Formel: C₁₀H₅BrO₃
Molecular Weight: 253,05 g/mol
MDL: MFCD00191849
CAS-Nummer: 52817-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR