2-(4-Methoxyphenoxy)benzaldehyd

Lieferant: BIONET RESEARCH

12N-027.500 12N-027.1
BIOT12N-027.500EA 0 EUR
BIOT12N-027.500 BIOT12N-027.1
2-(4-Methoxyphenoxy)benzaldehyd
2-(4-Methoxyphenoxy)benzaldehyde
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.2400 g/mol
MDL: MFCD01568732
CAS-Nummer: 19434-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR