4-(2,4-Dichlorphenoxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

661252-1G 661252-5G
SIAL661252-1GEA 118 EUR
SIAL661252-1G SIAL661252-5G
4-(2,4-Dichlorphenoxy)benzaldehyd, Sigma-Aldrich®
4-(2,4-Dichlorphenoxy)benzaldehyd
Formel: C₁₃H₈Cl₂O₂
MDL: MFCD08276810
CAS-Nummer: 78725-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR