3-(Benzyloxy)-4-methoxybenzaldehyd
Lieferant: Thermo Scientific
Synonyme:
3-Benzyloxy-4-methoxybenzaldehyde
TL00931.1
TL00931EA
TL00931EE
MAYBTL00931.1EA
0
EUR
MAYBTL00931.1
MAYBTL00931EA
MAYBTL00931EE
3-(Benzyloxy)-4-methoxybenzaldehyd
3-(Benzyloxy)-4-methoxybenzaldehyd
Formel:
C₁₅H₁₄O₃ Molecular Weight: 242,27 g/mol |
MDL:
MFCD00003386 CAS-Nummer: 6346-05-0 EINECS: 228-752-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...