Photographic trays
Lieferant: SEMADENI
0654 0655 0658
SEMA0654EA 71.4 EUR
SEMA0654 SEMA0655 SEMA0658
Photographic trays
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Trays made of PVC, inside white and outside grey in colour, with floor grooves.
  • Stable and cost effective
  • Resistant to acids
  • Temperature resistance up to 60 °C