2,3-Difluor-6-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
1,2-Difluoro-3-methoxy-4-nitrobenzene
AN-1347-5G
AN-1347-25G
AN-1347-100G
COBBAN-1347-5GEA
0
EUR
COBBAN-1347-5G
COBBAN-1347-25G
COBBAN-1347-100G
2,3-Difluor-6-nitroanisol
2,3-Difluor-6-nitroanisol
Formel:
C₇H₅F₂NO₃ |
MDL:
MFCD03840468 CAS-Nummer: 66684-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...