3,5-Difluorpicolinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3,5-Difluoropyridine-2-carboxylic acid
OS-7627-5G
OS-7627-1G
COBBOS-7627-5GEA
112
EUR
COBBOS-7627-5G
COBBOS-7627-1G
3,5-Difluorpicolinsäure
3,5-Difluorpicolinsäure
Formel:
C₆H₃F₂NO₂ Molecular Weight: 159.09 g/mol |
MDL:
MFCD05664124 CAS-Nummer: 745784-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...