Hexadecacarbonylhexarhodium ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hexarhodium hexadecacarbonyl
B24185.02
B24185.77
B24185.02EA
794
EUR
B24185.02
B24185.77
Hexadecacarbonylhexarhodium ≥98%
Hexadecacarbonylhexarhodium
Purity: 98%
CAS No.: 28407-51-4
Molecular Formula: Rh6(CO)16
Formula Weight: 1065.60
Melting Point:
Formel:
C₁₆O₁₆Rh₆ Molecular Weight: 1065.595 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011206 CAS-Nummer: 28407-51-4 EINECS: 249-009-3 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...