3-(4-Methylphenyl)-1H-pyrazol-5-amin

Lieferant: CHEMPUR

Synonyme: 5-Amino-3-(4-methylphenyl)pyrazole, 3-Amino-5-(4-methylphenyl)-1H-pyrazole, 3-Amino-5-p-Tolylpyrazole, 5-(p-Tolyl)-1H-pyrazol-3-amine

KM06993.250 KM06993.1
CHMPKM06993.250EA 0 EUR
CHMPKM06993.250 MAYBKM06993.1
3-(4-Methylphenyl)-1H-pyrazol-5-amin
3-(4-Methylphenyl)-1H-pyrazol-5-amin
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,2 g/mol
MDL: MFCD00084887
CAS-Nummer: 78597-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR