(S)-(-)-1-Boc-3-aminopyrrolidin 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

L19696.03 L19696.06
L19696.03EA 47.4 EUR
L19696.03 L19696.06
(S)-(-)-1-Boc-3-aminopyrrolidin 95%
(S)-(-)-1-Boc-3-aminopyrrolidin

Purity: 98% CAS No.: 147081-44-5 Molecular Formula: C9H18N2O2 Formula Weight: 186.25 Storage Temperature: 2°C to 8°C Refractive Index: 1.4715 Boiling Point: 115°/4mm Beilstein Reference: 8312125 Optical Rotation: -3° (neat) UN No.: 2735 MDL No.: MFCD03419271

Formel: C₉H₁₈N₂O₂
Molecular Weight: 186.25 g/mol
Siedepunkt: 115 °C (4 mmHg)
Dichte: 1.020
MDL: MFCD03419271
CAS-Nummer: 147081-44-5
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR