(S)-(-)-1-Boc-3-aminopyrrolidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

634794-1G 634794-5G
SIAL634794-1GEA 45.7 EUR
SIAL634794-1G SIAL634794-5G
(S)-(-)-1-Boc-3-aminopyrrolidin, Sigma-Aldrich®
(S)-(-)-1-Boc-3-aminopyrrolidin
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186.25 g/mol
MDL: MFCD03419271
CAS-Nummer: 147081-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR