(±)-1-Boc-3-aminopyrrolidin 96%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: 3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine
H29231.06 H29231.03
H29231.06EA 195 EUR
H29231.06 H29231.03
(±)-1-Boc-3-aminopyrrolidin 96%
(±)-1-Boc-3-aminopyrrolidin
Purity: 96% CAS No.: 186550-13-0 Molecular Formula: C9H18N2O2 Formula Weight: 186.25 Storage Temperature: 2°C to 8°C Refractive Index: 1.4710 UN No.: 2735 MDL No.: MFCD01861220
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186.25 g/mol
Dichte: 1.022
MDL: MFCD01861220
CAS-Nummer: 186550-13-0
ADR: 8 III