(±)-1-Boc-3-aminopyrrolidin

Lieferant: FLUOROCHEM

Synonyme: 3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine

011313-1G 011313-5G
FLUO011313-1GEA 60.9 EUR
FLUO011313-1G FLUO011313-5G
(±)-1-Boc-3-aminopyrrolidin
(±)-1-Boc-3-aminopyrrolidin
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186.25 g/mol
MDL: MFCD01861220
CAS-Nummer: 186550-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR