(±)-1-Boc-3-(methylamino)pyrrolidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 3-(methylamino)pyrrolidine-1-carboxylate

AM-1759-5G AM-1759-1G
COBBAM-1759-5GEA 221 EUR
COBBAM-1759-5G COBBAM-1759-1G
(±)-1-Boc-3-(methylamino)pyrrolidin
(±)-1-Boc-3-(methylamino)pyrrolidin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200.2780 g/mol
MDL: MFCD06656606
CAS-Nummer: 454712-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR