(R)-(-)-1-Boc-3-hydroxypyrrolidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(-)-N-Boc-3-pyrrolidinol, tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate, (R)-N-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine
M-Clarity™ Qualitätsstufe = MQ100
532169-1G
532169-5G
SIAL532169-1GEA
27
EUR
SIAL532169-1G
SIAL532169-5G
(R)-(-)-1-Boc-3-hydroxypyrrolidin, Sigma-Aldrich®
(R)-(-)-1-Boc-3-hydroxypyrrolidin
Formel:
C₉H₁₇NO₃ Molecular Weight: 187,24 g/mol |
MDL:
MFCD01317838 CAS-Nummer: 109431-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...