(S)-(+)-1-Boc-3-hydroxypyrrolidin
Lieferant: COMBI-BLOCKS
Synonyme:
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate
AM-1756-5G
AM-1756-25G
COBBAM-1756-5GEA
25.9
EUR
COBBAM-1756-5G
COBBAM-1756-25G
(S)-(+)-1-Boc-3-hydroxypyrrolidin
(S)-(+)-1-Boc-3-hydroxypyrrolidin
Formel:
C₉H₁₇NO₃ Molecular Weight: 187,2362 g/mol |
MDL:
MFCD01317839 CAS-Nummer: 101469-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...