(±)-1-Boc-3-hydroxypyrrolidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 3-hydroxypyrrolidine-1-carboxylate, 3-Hydroxypyrrolidine, N-BOC protected

OS-7537-25G OS-7537-5G OS-7537-1G
COBBOS-7537-25GEA 125 EUR
COBBOS-7537-25G COBBOS-7537-5G COBBOS-7537-1G
(±)-1-Boc-3-hydroxypyrrolidin
(±)-1-Boc-3-hydroxypyrrolidin
Formel: C₉H₁₇NO₃
Molecular Weight: 187,24 g/mol
MDL: MFCD04038535
CAS-Nummer: 103057-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR