2-Chlor-4-(trifluormethoxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Chloro-4-(trifluoromethoxy)phenyl)boronic acid, 2-Chloro-4-trifluoromethoxyphenylboronic acid
BB-2198-5G
BB-2198-1G
BB-2198-250M
COBBBB-2198-5GEA
2510
EUR
COBBBB-2198-5G
COBBBB-2198-1G
COBBBB-2198-250M
2-Chlor-4-(trifluormethoxy)phenylboronsäure
2-Chlor-4-(trifluormethoxy)phenylboronsäure
Formel:
C₇H₅BClF₃O₃ Molecular Weight: 240.3720 g/mol |
MDL:
MFCD11040272 CAS-Nummer: 345226-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...