Thallium(I)hexafluoracetylacetonat

Lieferant: FLUOROCHEM

Synonyme: Thallium(I) hexafluoro-2,4-pentanedionate

007416-1G 007416-5G
FLUO007416-1GEA 0 EUR
FLUO007416-1G FLUO007416-5G
Thallium(I)hexafluoracetylacetonat
Thallium(I)hexafluoracetylacetonat
Formel: C₁₅H₃F₁₈O₆Tl
Molecular Weight: 825,535 g/mol
MDL: MFCD00064761
CAS-Nummer: 15444-43-6
UN: 1707
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR