(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP) ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: (7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphonium hexafluorophosphate, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), PyAOP, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphorous hexafluorophosphate
H54378.03 H54378.06
H54378.03EA 93.8 EUR
H54378.03 H54378.06
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP) ≥99%
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
Purity: 99+% CAS No.: 156311-83-0 Molecular Formula: C17H27F6N7OP2 Formula Weight: 521.38 Melting Point: 163-168° MDL No.: MFCD03703417
Formel: C₁₇H₂₇F₆N₇OP₂
Schmelzpunkt: 163…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03703417
CAS-Nummer: 156311-83-0