(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)

Lieferant: BACHEM BIOCHEMICA

Synonyme: PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate

4031604.0025 4031604.0100
BACE4031604.0025EA 0 EUR
BACE4031604.0025 BACE4031604.0100
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
Formel: C₁₈H₂₈F₆N₆OP₂
Molecular Weight: 520,4 g/mol
MDL: MFCD00077411
CAS-Nummer: 128625-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR