(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
M-Clarity™ Qualitätsstufe = MQ200
377848-1G
377848-5G
SIAL377848-1GEA
29.6
EUR
SIAL377848-1G
SIAL377848-5G
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP), Sigma-Aldrich®
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
Formel:
C₁₈H₂₈F₆N₆OP₂ Molecular Weight: 520,4 g/mol |
MDL:
MFCD00077411 CAS-Nummer: 128625-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...