Hydrocortison-21-acetat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Hydrocortisone acetate

A18089.03 A18089.06
A18089.03EA 41.6 EUR
A18089.03 A18089.06
Hydrocortison-21-acetat ≥97%
Hydrocortison-21-acetat

Purity: 97+% CAS No.: 50-03-3 Molecular Formula: C23H32O6 Formula Weight: 404.51 Melting Point: ca 220° dec. Beilstein Reference: 2066841 Merck Reference: 14,4787 MDL No.: MFCD00037714

Formel: C₂₃H₃₂O₆
Molecular Weight: 404,51 g/mol
Schmelzpunkt: ∼ 220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037714
CAS-Nummer: 50-03-3
EINECS: 200-004-4
Merck Index: 14,4787

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR