Brefeldin A (natürlich)

Lieferant: Thermo Scientific

Synonyme: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural

297140250 297140050
ACRO297140250EA 428 EUR
ACRO297140250 ACRO297140050
Brefeldin A (natürlich)
Brefeldin A
Formel: C₁₆H₂₄O₄
Molecular Weight: 280,4 g/mol
Schmelzpunkt: 201…205 °C
Lagertemperatur: Kühlschrank
MDL: MFCD12913297
CAS-Nummer: 20350-15-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 201 to 205 °C
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR