Brefeldin A

Lieferant: Cayman Chemical

Synonyme: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural

11861-5 11861-10 11861-25
CAYM11861-5EA 46 EUR
CAYM11861-5 CAYM11861-10 CAYM11861-25
Brefeldin A
Brefeldin A
Formel: C₁₆H₂₄O₄
Molecular Weight: 280,4 g/mol
MDL: MFCD00083258
CAS-Nummer: 20350-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR