Brefeldin A, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
M-Clarity™ Qualitätsstufe = MQ300
B6542-25MG
B6542-5MG
SIALB6542-25MGEA
793
EUR
SIALB6542-25MG
SIALB6542-5MG
Brefeldin A, Sigma-Aldrich®
Brefeldin A
Formel:
C₁₆H₂₄O₄ Molecular Weight: 280,4 g/mol |
MDL:
MFCD00083258 CAS-Nummer: 20350-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...