D(+)-Glucose-[13C6,1,2,3,4,5,6,6-D7] (99% ¹³C , 97% D)
Lieferant: Merck
Synonyme:
D(+)-Glucose-[¹³C₆,1,2,3,4,5,6,6-²H₇]
M-Clarity™ Qualitätsstufe = MQ200
552151-1G
552151-5G
SIAL552151-1GEA
982
EUR
SIAL552151-1G
SIAL552151-5G
D(+)-Glucose-[13C6,1,2,3,4,5,6,6-D7] (99% ¹³C , 97% D)
D(+)-Glucose-[13C6,1,2,3,4,5,6,6-D7]
Formel:
C₆H₅D₇O₆ |
MDL:
MFCD00274634 CAS-Nummer: 201417-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...