1-(4-Bromphenylsulfonyl)piperazin

Lieferant: COMBI-BLOCKS

OT-0925-5G OT-0925-25G OT-0925-1G
COBBOT-0925-5GEA 98.3 EUR
COBBOT-0925-5G COBBOT-0925-25G COBBOT-0925-1G
1-(4-Bromphenylsulfonyl)piperazin
1-(4-Bromphenylsulfonyl)piperazin
Formel: C₁₀H₁₃BrN₂O₂S
MDL: MFCD00474647
CAS-Nummer: 179334-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR