(±)-2-Piperazincarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Piperazine-2-carboxylic acid
AM-1754-5G
AM-1754-25G
AM-1754-1G
COBBAM-1754-5GEA
25.9
EUR
COBBAM-1754-5G
COBBAM-1754-25G
COBBAM-1754-1G
(±)-2-Piperazincarbonsäure
(±)-2-Piperazincarbonsäure
Formel:
C₅H₁₀N₂O₂ Molecular Weight: 130,15 g/mol |
MDL:
MFCD00132878 CAS-Nummer: 2762-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...