1-Boc-piperazin-2-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

AM-1744-5G AM-1744-1G
COBBAM-1744-5GEA 324 EUR
COBBAM-1744-5G COBBAM-1744-1G
1-Boc-piperazin-2-carbonsäure
1-Boc-piperazin-2-carbonsäure
Formel: C₁₀H₁₈N₂O₄
Molecular Weight: 230,2600 g/mol
MDL: MFCD02682403
CAS-Nummer: 1214196-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR