(±)-2-Piperazincarbonsäure Dihydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Piperazine-2-carboxylic acid dihydrochloride

133660100 AC13366.50 AC13366DA 133660500
ACRO133660100EA 76.7 EUR
ACRO133660100 MAYBAC13366.50 MAYBAC13366DA ACRO133660500
(±)-2-Piperazincarbonsäure Dihydrochlorid 98%
(±)-2-Piperazincarbonsäure Dihydrochlorid
Formel: C₅H₁₂Cl₂N₂O₂
Molecular Weight: 203.07 g/mol
Schmelzpunkt: 200 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00044803
CAS-Nummer: 3022-15-9

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Titration with HClO4 97.5 to 102.5 %
NMR (31P-NMR) Consistent with structure
Specific optical rotation +35° to +45° (20°C, 589 nm) (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR