(±)-2-Piperazincarbonsäure Dihydrochlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Piperazine-2-carboxylic acid dihydrochloride
H27101.14
H27101.06
H27101.14EA
102
EUR
H27101.14
H27101.06
(±)-2-Piperazincarbonsäure Dihydrochlorid ≥98%
(±)-2-Piperazincarbonsäure Dihydrochlorid
Purity: 98%
CAS No.: 3022-15-9
Molecular Formula: C5H10N2O2.2HCl
Formula Weight: 203.07
Melting Point: 270-273°
MDL No.: MFCD00044803
Formel:
C₅H₁₂Cl₂N₂O₂ Molecular Weight: 203,07 g/mol Schmelzpunkt: 270…273 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044803 CAS-Nummer: 3022-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...