1-(Ethylsulfonyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

653306-1G 653306-5G
SIAL653306-1GEA 62.1 EUR
SIAL653306-1G SIAL653306-5G
1-(Ethylsulfonyl)piperazin, Sigma-Aldrich®
1-(Ethylsulfonyl)piperazin
Formel: C₆H₁₄N₂O₂S
Molecular Weight: 178,25 g/mol
MDL: MFCD02093528
CAS-Nummer: 62937-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR