1-(Methylsulfonyl)piperazin

Lieferant: FLUOROCHEM

Synonyme: 1-Methanesulphonylpiperazine

022119-5G 022119-1G
FLUO022119-5GEA 113 EUR
FLUO022119-5G FLUO022119-1G
1-(Methylsulfonyl)piperazin
1-(Methylsulfonyl)piperazin
Formel: C₅H₁₂N₂O₂S
Molecular Weight: 164,23 g/mol
MDL: MFCD02153225
CAS-Nummer: 55276-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR