1-(4-Bromphenylsulfonyl)-4-isobutylpiperazin

Lieferant: COMBI-BLOCKS

OT-0972-5G OT-0972-1G
COBBOT-0972-5GEA 303 EUR
COBBOT-0972-5G COBBOT-0972-1G
1-(4-Bromphenylsulfonyl)-4-isobutylpiperazin
1-(4-Bromphenylsulfonyl)-4-isobutylpiperazin
Formel: C₁₄H₂₁BrN₂O₂S
MDL: MFCD12913974
CAS-Nummer: 1187385-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR