1-(4-Bromphenylsulfonyl)-4-butylpiperazin

Lieferant: COMBI-BLOCKS

OT-0946-5G OT-0946-1G
COBBOT-0946-5GEA 303 EUR
COBBOT-0946-5G COBBOT-0946-1G
1-(4-Bromphenylsulfonyl)-4-butylpiperazin
1-(4-Bromphenylsulfonyl)-4-butylpiperazin
Formel: C₁₄H₂₁BrN₂O₂S
MDL: MFCD13195730
CAS-Nummer: 1199773-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR