1-(4-Bromphenylsulfonyl)-4-ethylpiperazin

Lieferant: COMBI-BLOCKS

OT-0943-5G OT-0943-1G
COBBOT-0943-5GEA 303 EUR
COBBOT-0943-5G COBBOT-0943-1G
1-(4-Bromphenylsulfonyl)-4-ethylpiperazin
1-(4-Bromphenylsulfonyl)-4-ethylpiperazin
Formel: C₁₂H₁₇BrN₂O₂S
MDL: MFCD02656010
CAS-Nummer: 385404-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR