1-(4-Bromphenylsulfonyl)-4-methylpiperazin
Lieferant: COMBI-BLOCKS
Synonyme:
1-((4-Bromophenyl)sulphonyl)-4-methylpiperazine
OT-0945-5G
OT-0945-1G
COBBOT-0945-5GEA
451
EUR
COBBOT-0945-5G
COBBOT-0945-1G
1-(4-Bromphenylsulfonyl)-4-methylpiperazin
1-(4-Bromphenylsulfonyl)-4-methylpiperazin
Formel:
C₁₁H₁₅BrN₂O₂S Molecular Weight: 319.2180 g/mol |
MDL:
MFCD00595522 CAS-Nummer: 837-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...